Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Transfertube™ Tube
SDP

Catalog No. 0880125 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Capacity (Metric):
4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
08-801-25 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-801-25 Supplier Spectrum™ Supplier No. 190195
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For transferring and coring liquid and solid samples

  • Material: polypropylene
  • Volume: 4mL
  • 153mm length × 6mm dia.
  • I.D.: 16.5mm
  • O.D.: 20mm

Specifications

Capacity (Metric) 4 mL
Length (Metric) 175 mm
Material Polypropylene
For Use With Bacterial or viral colony on agar in microbiology, powders, viscous solutions, slurries, liquids, soil, mud, sludge, core samples
Diameter (Metric) 20 mm
Product Type Transport Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.