Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Spectrum Tube Power Supply, 220 V Euro Plug
SCI_ED

Catalog No. S27617 Shop All Eisco Products
Change view
Click to view available options
Grade:
10 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27617 10 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27617 Supplier Eisco™ Supplier No. ESRPH1197A220EU
Add to Cart
Add to Cart

This spectrum tube power supply is used to operate spectrum tubes safely in the classroom or laboratory with unique safety features.

  • European Style Plug, 220 V
  • Flat black finish to help eliminate ambient light
  • Unique casing design for holding 26 cm spectrum tubes securely in place
  • Prevents users from touching electrodes
  • On-off toggle switch for ease of use
  • CE, CSA and CUS approved
  • Safety door
  • Spectrum tubes sold separately

Specifications

Certifications/Compliance CE, CSA and CUS Approved
Color Black
Grade 10 to Undergraduate
Age 14 to 20
Electrical Requirements 220 V
For Use With (Application) Safely Operating Spectrum Tubes
Length (Metric) 30 cm
Product Type Power Supply
Description European Plug
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.