Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Spectrum Tubes
SDPSCI_ED

Catalog No. S07000 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type:
Air Spectrum Tube
Argon Spectrum Tube
Carbon-di-Oxide Spectrum Tube
Helium Spectrum Tube
Hydrogen Spectrum Tube
Krypton Spectrum Tube
Neon Spectrum Tube
Nitrogen Spectrum Tube
Oxygen Spectrum Tube
Water vapor Spectrum Tube
Xenon Spectrum Tube
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S07000 Carbon-di-Oxide Spectrum Tube
S06995 Helium Spectrum Tube
S06996 Neon Spectrum Tube
S06997 Oxygen Spectrum Tube
S06994 Hydrogen Spectrum Tube
S06998 Water vapor Spectrum Tube
S06999 Air Spectrum Tube
S07006 Argon Spectrum Tube
S07004 Xenon Spectrum Tube
S07002 Nitrogen Spectrum Tube
S07003 Krypton Spectrum Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. S07000 Supplier United Scientific™ Supplier No. SPTCD01
Only null left
Add to Cart
Add to Cart

The classic demonstration of spectral lines.

Acquaint students with the spectra of common gases in their pure form while demonstrating concept of wavelength.

  • 25 cm (10 in.) long
  • Capillary portion about 4 in. (10 cm) long

Specifications

Grade 5 to 12
For Use With (Application) Emits characteristic spectrum when energized by spectroscope
For Use With (Equipment) Spectrometer
Includes Sealed electrode, metal endcap, loop and activity guide
Length (English) 10 in.
Length (Metric) 25.4 cm
Length (English) Capillary 4 in.
Length (Metric) Capillary 10.16 cm
Product Type Carbon-di-Oxide Spectrum Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.