Learn More
Sigma Aldrich Fine Chemicals Biosciences Rigid PTfE sleeve Joint S1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z2154301EA
Robust PTfE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free high-vacuum performance at temperatures up to 280C.