Learn More
World Precision Instrument Wire Retractor, 5cm, 10x5mm Blades, German
SDP
Supplier:  World Precision Instrument 14130G
Encompass_Preferred
A wire eyelid speculum is a surgical instrument used by ophthalmologists to keep eyelids open during an exam or surgery. It consists of two curved blades that are gently inserted between the upper and lower eyelids to hold them open, allowing the ophthalmologist a clear view of the eye. Our small wire speculum is made of surgical grade stainless steel or our premium German steel, and it comes in two sizes, with a spread of 13 and 25 mm. This wire speculum can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.
Catalog No. 50-822-247
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.