Learn More
Description
- Maintenance free and easy to install speed bump
- Channeled bottom covers wires and cabling
- Recycled rubber composition ensures parking stops never chip, crack, nor
- require painting
- Rubber composition keeps parking stop from ever needing painting
- Able to withstand and endure abuse from gas, oil, salt, sunlight, and chemicals
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Speed Bump |
| Packaging Quantity | 1 Ea. |
| Material | Plastic |
| Type | End Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.