Learn More
3M™ Speedglas™ 9100 Welding Helmet Outside Protection Plate, High Temperature
Catalog No. 19070956 Shop All 3M Products
Change view
Click to view available options
Welding Helmet Replacement Part: Protection Plate
Catalog No.
19-070-956 Welding Helmet Replacement Part: Protection Plate
1 options
Catalog No. 19-070-956 Supplier 3M™ Supplier No. 7000127128
Add to Cart
Add to Cart
The Speedglas™ 9100 high temperature outside protection plate is formulated from an engineered resin to resist higher heat applications. This is recommended for use in high amperage welding or conditions where long weld beads may generate radiant heat.
  • Outside Protection Plate
  • High Temp Applications
  • Replacement Part
  • 3M Catalog# 06-0200-53
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010
For Use With (Equipment) Speedglas 9100 Helmets
For Use With (Application) Facility Maintenance, Assembly, Welding, Sanding, Machine Operations, Grinding
Lens Tint None
Model 06-0200-53
Product Type High-temperature Outside Protection Plate
Quantity 10/Pk.