Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ APF Kit with Flame Retardant Neck Shroud and Large Head Cover

Catalog No. 19072133 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-133 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-133 Supplier 3M™ Supplier No. 7100216088
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ APF Kit is for use with the complete helmet assembly to help protect the user from welding sparks, spatter, and radiation on the ears and neck.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 3M Speedglas Heavy-duty Welding Helmet G5-01
For Use With (Application) Metal Processing, Grinding, Metal Repair, Welding
Model 46-1000-00
Product Type APF Kit
Quantity 1 Ea.
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.