Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Battery Holder

Catalog No. 19072173 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M Speedglas Heavy-duty Welding Helmet G5-01
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-072-173 3M Speedglas Heavy-duty Welding Helmet G5-01
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-173 Supplier 3M™ Supplier No. 7100214393
Add to Cart
Add to Cart

Spare part to the Speedglas welding filter series G5-01.

  • For use if the original battery holder becomes worn or damaged during use.
  • For maximum safety and comfort, make it a habit to check equipment regularly.
  • Replace worn out or damaged parts.
    TRUSTED_SUSTAINABILITY

    Specifications

    Color Black
    For Use With (Equipment) 3M Speedglas Heavy-duty Welding Helmet G5-01
    For Use With (Application) Metal Processing, Welding, Grinding, Metal Repair
    Model 44-0400-55
    Product Type Battery Holder
    Quantity 1 Ea.
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.