Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Cotton Sweatband

Catalog No. 19072145 Shop All 3M Products
Change view
Click to view available options
Material:
Cotton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-072-145 Cotton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-145 Supplier 3M™ Supplier No. 7100232455
Add to Cart
Add to Cart

Can be utilized with welding helmet series 9100 and G5-01.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 3M Speedglas Heavy-duty Welding Helmet G5-01
For Use With (Application) Metal Processing, Welding, Metal Repair, Grinding
Material Cotton
Model 46-0200-54
Product Type Sweatband
Quantity 3/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.