Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Fabric Head Cover

Catalog No. 19072131 Shop All 3M Products
Change view
Click to view available options
Size:
Large
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-072-131 Large
19-072-130 Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-131 Supplier 3M™ Supplier No. 7100215555
Add to Cart
Add to Cart

3M™ Speedglas™ Fabric Head Cover is made in flame-retardant fabric for protection against sparks and spatter.

  • A variety of accessories are available for a Speedglas welding helmet to help maximize protection and comfort
  • For maximum safety and comfort, make it a habit to check equipment regularly
  • Replace worn out or damaged parts.
  • Large head coverage option is for use with the complete helmet assembly to help protect the user from welding sparks and spatter on the top portion of the head.
TRUSTED_SUSTAINABILITY

Specifications

Construction Flame retardant
Color Black
For Use With (Equipment) Speedglas Heavy-duty Welding Helmet G5-01
For Use With (Application) Grinding, Welding, Metal Processing, Metal Repair
Model 46-0700-81
Product Type Headcover
Quantity 1 Ea.
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.