Learn More
3M™ Speedglas™ Head Suspension Assembly
Catalog No. 19072142 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M Speedglas Heavy-duty Welding Helmet G5-01
Catalog No. For Use With (Equipment)
19-072-142 3M Speedglas Heavy-duty Welding Helmet G5-01
1 options
Catalog No. 19-072-142 Supplier 3M™ Supplier No. 7100259734
Add to Cart
Add to Cart
Highly adjustable, comfortable, headband for the 3M™ Speedglas™ Heavy-Duty Welding Helmet G5-01.
  • Smooth ratchet for precise tightening, fits head sizes 50 to 64.
  • Includes large comfort head rest.
  • For maximum safety and comfort, make it a habit to check equipment regularly.
    TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) 3M Speedglas Heavy-duty Welding Helmet G5-01
    For Use With (Application) Metal Processing, Grinding, Metal Repair, Welding
    Includes Large comfort head rest
    Product Type Suspension Assembly
    Quantity 1 Ea.