Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Heavy-Duty Welding Helmet with Adflo™ High-Altitude PAPR Assembly, No ADF

Catalog No. 19072122 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-122 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-122 Supplier 3M™ Supplier No. 7100214700

missing translation for 'validMainframeMsg'

3M™ Speedglas™ Heavy-Duty Welding Helmet features a slim profile and ergonomic design that makes it more lightweight and easier to utilize in tight welding spaces

  • Big (104 x 170 mm), clear protective visor is curved, with excellent views up, down and peripherally.
  • For more control over comfort, the innovative G5-01 helmet airflow optimization system lets users direct the airflow to either theface, visor or somewhere in-between.
  • Re-configure helmet using many of the additional available accessories to find the right coverage and protection. (Optional accessories include: task light assembly, 1000 Assigned Protection Factor (APF) assembly, high-visibility head cover, leather head/neck cover, and more.
  • With the right type of filter and/or cartridge, the Adflo respirator effectively protects users against both particles and gases - all in one system.
  • NIOSH approved and provides an Assigned Protection Factor (APF) of 25 in the standard helmet assembly, and up to 1000 APF with the optional kit.
  • G5-01 ADFs are Bluetooth enabled meaning users get access to additional functionalities, statistics and documentation.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI Z87.1-2015; NIOSH Approved
Color Black
For Use With (Equipment) 3M Adflo Cartridge
For Use With (Application) Grinding, Welding, Metal Processing, Metal Repair
Includes Adflo™ High-Altitude PAPR Assembly
Model 46-1101-00
Product Type Welding Respiratory System
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.