Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Heavy-Duty Welding Helmet, Rigid Neck Cover, Fabric Head Cover, No ADF

Catalog No. 19072156 Shop All 3M Products
Change view
Click to view available options
Material:
Polyphthalamide (PPA)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-072-156 Polyphthalamide (PPA)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-156 Supplier 3M™ Supplier No. 7100259574
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Speedglas™ Heavy-Duty Welding Helmet features a slim profile and ergonomic design that makes it more lightweight and easier to utilize in tight welding spaces .

  • The big (104 x 170 mm), clear protective visor is curved, with excellent views up, down and peripherally.
  • For more control over comfort, the innovative G5-01 helmet airflow optimization system lets users direct the airflow to either the face, visor or somewhere in-between.
    TRUSTED_SUSTAINABILITY

    Specifications

    Color Black
    For Use With (Application) Grinding, Metal Repair, Welding, Metal Processing
    Material Polyphthalamide (PPA)
    Model 46-0099-35
    Product Type Welding Helmet
    Quantity 1 Ea.
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.