Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Leather Neck Cover

Catalog No. 19072165 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-165 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-165 Supplier 3M™ Supplier No. 7100215744
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Speedglas™ Leather Neck Cover is made in rugged leather for more long term protection against sparks and spatter.

  • For maximum safety and comfort, make it a habit to check equipment regularly.
  • Replace worn out or damaged parts.
    TRUSTED_SUSTAINABILITY

    Specifications

    Construction Flame retardant
    Color Brown
    For Use With (Application) Metal Repair, Metal Processing, Grinding, Welding
    Model 46-0700-67
    Product Type Neck Cover
    Quantity 1 Ea.
    Visor Type Clear Visor and Auto-Darkening Welding Lens
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.