Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Long Battery Power Cable for Task Light

Catalog No. 19072147 Shop All 3M Products
Change view
Click to view available options
Quantity:
1/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-147 1/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-147 Supplier 3M™ Supplier No. 7100216920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

G5-01 Long Battery Power Cable for Task Light.

  • For maximum safety and comfort, make it a habit to check your equipment regularly.
  • Replace worn out or damaged parts.
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) Speedglas Heavy-duty Welding Helmet G5-01 Task Light
For Use With (Application) Grinding, Metal Processing, Metal Repair, Welding
Model 46-0500-02
Product Type Power Cable
Quantity 1/Bag
Size Long
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.