Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Speedglas™ Task Light

Catalog No. 19072126 Shop All 3M Products
Change view
Click to view available options
Quantity:
1/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-072-126 1/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-126 Supplier 3M™ Supplier No. 7100217182

missing translation for 'validMainframeMsg'

Used if the original task light becomes worn or damaged

  • No power cables or protection plates included
  • For maximum safety and comfort, make it a habit to check equipment regularly.
  • Replace worn out or damaged parts.
    TRUSTED_SUSTAINABILITY

    Specifications

    Color Black
    For Use With (Equipment) 3M Speedglas Heavy-duty Welding Helmet G5-01
    For Use With (Application) Metal Processing, Metal Repair, Welding, Grinding
    Model 46-0500-05
    Product Type Light
    Quantity 1/Bag
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.