Learn More
3M™ Speedglas™ Welding Helmet Magnification Plate
Catalog No. 19070955 Shop All 3M Products
Change view
Click to view available options
Product Type:
Magnifying Lens 2.0X
Magnifying Lens 3.0X
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-070-955 Magnifying Lens 3.0X
19-070-954 Magnifying Lens 2.0X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-070-955 Supplier 3M™ Supplier No. 7000127146
Add to Cart
Add to Cart
The magnifier is used for magnifying objects while inserted into the Speedglas™ 9100 ADF on the inside slot.
  • Magnification Plate
  • Compatible with 9100 Series ADFs
  • 3M Catalog# 06-0500-57 , 3M Catalog# 06-0500-59
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z87.1-2010
For Use With (Equipment) Speedglas 9100 ADF
For Use With (Application) Welding, Sanding, Grinding, Facility Maintenance, Machine Operations, Assembly
Lens Tint None
Model 06-0500-59
Product Type Magnifying Lens 3.0X
Quantity 1 Ea.