Learn More
Description
- 3M Catalog# 06-0005-P10, 06-0005-P11, 06-0005-P12, 06-0005-P13
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | 9100 Series Welding Helmets |
| For Use With (Application) | Facility Maintenance, Grinding, Machine Operations, Sanding, Welding |
| Lens Tint | Passive shade 12 |
| Model | 06-0005-P12 |
| Product Type | Welding Lens |
| Quantity | 2/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.