Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator and Rotors

Catalog No. SRF110230
Change view
Click to view available options
Type:
12-13 mm Rotor
12-17 mm Rotor
28 mm Rotor
Hexagonal Microcentrifuge Rotor
Microplate Rotor
Vacuum Concentrator
Vacuum Concentrator Kit
Vacuum Concentrator Universal Vacuum Kit
Electrical Requirements:
115 V, 60 Hz, 16 A
230 V, 50 Hz, 8 A
N/A
Includes:
Concentrator
Oil Pump, Vapor Trap, Accessories
Rotor
Universal Vacuum System, Rotor, Accessories
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Electrical Requirements Includes
SRF110230 Vacuum Concentrator 230 V, 50 Hz, 8 A Concentrator
RF108 12-13 mm Rotor N/A Rotor
RF132 Hexagonal Microcentrifuge Rotor N/A Rotor
RF32 28 mm Rotor N/A Rotor
RF52 12-17 mm Rotor N/A Rotor
RFMT4 Microplate Rotor N/A Rotor
SRF110115 Vacuum Concentrator 115 V, 60 Hz, 16 A Concentrator
SRF110P1115 Vacuum Concentrator Kit 115 V, 60 Hz, 16 A Oil Pump, Vapor Trap, Accessories
SRF110P1230 Vacuum Concentrator Kit 230 V, 50 Hz, 8 A Oil Pump, Vapor Trap, Accessories
SRF110P2115 Vacuum Concentrator Universal Vacuum Kit 115 V, 60 Hz, 16 A Universal Vacuum System, Rotor, Accessories
SRF110P2230 Vacuum Concentrator Universal Vacuum Kit 230 V, 50 Hz, 8 A Universal Vacuum System, Rotor, Accessories
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. SRF110230 Supplier Thermo Scientific™ Supplier No. SRF110230
Add to Cart
Add to Cart
Request a Quote
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Process heat sensitive biological samples with confidence using SRF110. This Refrigerated SpeedVac system uses state-of-the-art centrifugation, vacuum, and heat technologies for removing solvents and concentrating heat sensitive biological samples while maintaining sample integrity.

SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules. They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.

SRF110 modular refrigerated vacuum concentrator provides flexibility to any benchtop space.  SRF110 can be configured depending on your applications need. Select from wide range of suitable ThermoFisher Scientific components such as pumps, Vapor Trap, and additional accessories to configure your custom vacuum concentration setup.  For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.

  • SRF110 offers unique programming capability to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules
  • Ideal for evaporation of non-aggressive, aqueous based solvents, buffers, HPLC fractions, etc.
  • 3 Preset program with option to store up to 9 programs
  • Temperature range -4°C up to 100°C (1°C increment)
  • Self aligning and quick release rotor mechanism
  • Hexagonal rotor (RF132) included with purchase of SRF110
  • Refer to list of rotors available for purchase and to be used with SRF110 model
  • Electricals- 115V AC (60Hz, 16Amps) and 230V AC (50Hz, 8Amps)
  • Certifications- UL Certified, CE, UKCA, KC, RCM Marked.
  • F Gas Compliance- Yes (EPA-SNAP and EU)
  • SRF110P1 kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), an Oil pump (VLP120-115), a vapor Trap (RVT450-115), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
  • SRF110P2 kit includes SRF110P2 Kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), Universal Vacuum System (UVS450), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)

Warranty and Services

1 year

TRUSTED_SUSTAINABILITY

Specifications

Includes Concentrator
Type Vacuum Concentrator
Shipping Weight (English) 108 lb.
Shipping Weight (Metric) 49 kg
Certifications CE, UKCA, KC, RCM
Temperature Range (Metric) -4°C to +100°C
Chamber Material Coated Aluminum
Description Medium Capacity, Modular Centrifugal Vacuum Concentrator
Electrical Requirements 230 V, 50 Hz, 8 A
Material Powder-coated Steel
Weight (English) 95 lb.
Weight (Metric) 43 kg
Dimensions (L x W x H) 56 x 37 x 43 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.