Learn More
Thermo Scientific™ SpeedVac™ SRF110 Refrigerated Centrifugal Vacuum Concentrator and Rotors
Description
SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility to your lab. It offers unique programming capabilities to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules. They are ideal for evaporation of aqueous based solvents, buffers, and HPLC fractions. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
SRF110 modular refrigerated vacuum concentrator provides flexibility to any benchtop space. SRF110 can be configured depending on your applications need. Select from wide range of suitable ThermoFisher Scientific components such as pumps, Vapor Trap, and additional accessories to configure your custom vacuum concentration setup. For a convenient, one model shopping experience select from SRF110P1 (modular configuration) and SRF110P2 (Semi integrated configuration) kit options. Each kit is designed to include components required for convenient SpeedVac setup.
- SRF110 offers unique programming capability to maintain chamber temperature pre and post sample run to prevent thermal degradation of heat sensitive samples such as proteins, enzymes, and other biomolecules
- Ideal for evaporation of non-aggressive, aqueous based solvents, buffers, HPLC fractions, etc.
- 3 Preset program with option to store up to 9 programs
- Temperature range -4°C up to 100°C (1°C increment)
- Self aligning and quick release rotor mechanism
- Hexagonal rotor (RF132) included with purchase of SRF110
- Refer to list of rotors available for purchase and to be used with SRF110 model
- Electricals- 115V AC (60Hz, 16Amps) and 230V AC (50Hz, 8Amps)
- Certifications- UL Certified, CE, UKCA, KC, RCM Marked.
- F Gas Compliance- Yes (EPA-SNAP and EU)
- SRF110P1 kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), an Oil pump (VLP120-115), a vapor Trap (RVT450-115), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
- SRF110P2 kit includes SRF110P2 Kit includes Refrigerated SpeedVac (SRF110), Hexagonal rotor (RF132), Universal Vacuum System (UVS450), Universal Tubing (UTP-TYG), 4L capacity Glass Condensation Flask (GCF400), Glass Cap (FC400), and 1L Bottle of heat transfer fluid Cryocool™ (SCC1)
Warranty and Services
1 year
Specifications
Specifications
| Type | Vacuum Concentrator Kit |
| Includes | Oil Pump, Vapor Trap, Accessories |
| Shipping Weight (English) | 108 lb. |
| Shipping Weight (Metric) | 49 kg |
| Certifications | UL, EPA-SNAP Fgas |
| Temperature Range (Metric) | -4°C to +100°C |
| Chamber Material | Coated Aluminum |
| Description | Medium capacity centrifugal vacuum concentrator kit with oil Pump, refrigerated vapor trap, rotor, accessories |
| Electrical Requirements | 115 V, 60 Hz, 16 A |
| Material | Powder-coated Steel |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.