Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPEF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121470 Supplier Invitrogen™ Supplier No. PA5121470
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody tested in Indirect ELISA.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SPEF2
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 2.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SPEF2
Gene Accession No. Q9C093
Gene Alias C230086A09Rik; cancer/testis antigen 122; CT122; KIAA1770; Kpl2; Protein KPL2; SPEF2; sperm flagellar 2; sperm flagellar protein 2; testis tissue sperm-binding protein Li 47a
Gene Symbols SPEF2
Host Species Rabbit
Immunogen Recombinant Human Sperm flagellar protein 2 protein (932-1230aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79925
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.