Learn More
Honeywell™ Sperian Safety Nuisance Disposable Dust Mask
Catalog No. 19005184 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Packaging Quantity:
50/Pk.
Catalog No. Packaging Quantity
19-005-184 50/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 19-005-184 Supplier Honeywell™ Supplier No. 14110094CC
Add to Cart
Add to Cart
This item is not returnable. View return policy
Scientifically designed for better comfort and protection
  • Respiratory protection from dirt and dust
  • Highly affordable disposable respirator
  • Defends against non-toxic mold and pollen
  • Recommended for agriculture, construction, and forestry
  • Great for everyday sweeping and dusting
  • Adjustable nose bridge clip offers a secure fit
  • Disposable, not NIOSH approved
Specifications
Product Type General Purpose Face Mask
Color White
Size One Size
Disposable Single-use
For Use With (Application) Provides basic protection against non toxic dust, pollen, mold and dander. Recommended for the agriculture, construction, and forestry industries. Also perfect for day-to-day dusting and sweeping, DIY projects, yard work, and other applications with non-toxic mold, pollen, dust and dander.
Latex Free Yes
Packaging Quantity 50/Pk.