Learn More
Honeywell™ SAF-T-FIT™ N99 Disposable Particulate Respirator
Catalog No. 19034911
Change view
Click to view available options
Size:
Small
X-Large
With Exhalation Valve:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size With Exhalation Valve
19-034-911 Small No
19-034-912 X-Large Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-034-911 Supplier Honeywell™ Supplier No. 14110402
This item is not returnable. View return policy
N99 protection combined with a full sealing flange for enhanced and comfortable fit.
  • Adjustable straps for a snug fit
  • Exhalation valve for ease of breathing during extended wear
  • Boomerang nose seal is adjustable for added comfort and a secure fit
  • Nose clip is color coded for visible size recognition
  • NIOSH Approval TC-84A-3845
  • Latex-free
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N99, approval number: TC-84A-3845
Color Code White
Design Type Cup/Molded
Disposable Single-use
Filter Type N99
Product Type General Purpose Particulate Respirator
Size Small
With Exhalation Valve No
Filter Efficiency 99%
For Use With (Application) Welding, soldering, metal cutting and pouring, torch cutting, grinding, sanding, sweeping, hot temperatures, general industrial applications and dusty operation
Latex Free Yes
Model 14110402
Nose Piece Material Metal
Nosepiece Type Nose Clip
Quantity 10/Pk.
Strap Attachment Type Dual Adjustable Straps
FDA Cleared Surgical Mask No