Learn More
Honeywell™ Sperian™ Survivair CF-SAR System Plugs
Catalog No. 17377774 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Connection Type:
Foster
Catalog No. Connection Type
17-377-774 Foster
1 options
Catalog No. 17-377-774 Supplier Honeywell™ Supplier No. 930429
Add to Cart
Add to Cart
Hansen and foster plugs for use with Sperian supplied air respirators.
Connects the SAR Attachment Kit (all versions) to an airline hose with compatible couplings. Available with Foster, Schrader and Hansen-compatible plugs.
Specifications
Connection Type Foster
Product Type Plug
For Use With (Application) Connects the SAR attachment kit (all versions) to an airline hose
For Use With (Equipment) Constant flow supplied air system
Model 930429