Learn More
Honeywell™ Sperian™ Survivair T-Series Cartridges
Description
- Cartridges designed to protect against select organic vapors and acid gases
- Selection is dependent on type of contaminant, level of exposure, and length of exposure
- Twist-Lock Cartridges provide an audible click indicating a positive lock on connector
- 6 cartridges per pack
Specifications
Specifications
| Cartridge Type | Organic Vapor, Acid Gas |
| Certifications (NIOSH) | NIOSH 42 CFR 84, TC-23C-741 |
| Product Type | Cartridge |
| For Use With (Equipment) | Sperian™ T-Series Facepieces |
| Quantity | 6/Pk. |