Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Sperian™ Waste Water Cart

Catalog No. 172923L Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-292-3L Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-292-3L Supplier Honeywell™ Supplier No. 320005110000
  • $282.70
    $227.65 / Each of 1

    Save $55.05

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Fendall Porta Stream I, II, III or Fendall Flash Flood

Allows easy collection of used eyewash flushing fluid

  • For use with Fendall Porta Stream I, II, II and Fendall Flash Flood
  • 23.5 gal. waste container cart allows easy collection of used flushing fluid
  • Concave top surface with center drain
  • Dual wheels
  • Molded handle for easy portability
  • Color: Green

Specifications

For Use With (Equipment) Porta Stream I, II, III and Flash Flood EyeWash
Height (English) 33.75 in.
Height (Metric) 85.7 cm
Includes Center Drain, Dual Wheels, Molded Handle
Material Polyethylene
Product Type Fluid Disposal Cart
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.