Learn More
Walter Products™ Spermatogenesis Model
SCI_ED
Catalog No. S139166 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W x H):
21 x 15 x 2 in.
Catalog No. Dimensions (L x W x H)
S139166 21 x 15 x 2 in.
1 options
This item is not returnable. View return policy
Catalog No. S139166 Supplier Walter Products™ Supplier No. B10503
Add to Cart
Add to Cart
This item is not returnable. View return policy
Model depicts the stages of spermatogenesis enlarged approximately 10,000 times.
  • Magnified process of spermatogenesis from spermatid to the mature spermatozoon is shown
  • All significant features are numbered and identifiable on the included key
  • Mounted on a durable plastic base
Specifications
Dimensions (L x W x H) 21 x 15 x 2 in.
Material PVC
Product Type Anatomical Model