Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Spermidine Hydrochloride, nNOS Inhibitor, Solid, 100 mg

Catalog No. 502707898
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7898 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7898 Supplier Abcam Supplier No. AB120057100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Endogenous polyamine that inhibits neuronal nitric oxide synthase and binds to the NMDA receptor.

  • Molecular Weight: 254.6 Da
  • Function: Endogenous polyamine that binds to the polyamine modulatory site of the NMDA receptor and inhibits neuronal nitric oxide synthase (nNOS)
  • Application: Binds and precipitates DNA; may be used for purification of DNA binding proteins
  • Form: Solid
  • Composition: 100% N-(3-aminopropyl)butane-1,4-diamine
  • Storage: Store at +4°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% N-(3-aminopropyl)butane-1,4-diamine
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Content And Storage Store at 4°C.
Substrate DNA
Target Molecule NMDA Receptor, Neuronal Nitric Oxide Synthase, DNA
Formulation 100% N-(3-aminopropyl)butane-1,4-diamine
Quantity 100 mg
For Use With (Application) nNOS inhibition, NMDA receptor research, DNA binding protein purification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.