Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermine, free base, ≥97%, MP Biomedicals™

Catalog No. ICN15207001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15207001 1 g
ICN15207005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15207001 Supplier MP Biomedicals, Inc Supplier No. 0215207001
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71-44-3
Molecular Formula C10H26N4
Molecular Weight (g/mol) 202.35
MDL Number MFCD00008215
InChI Key PFNFFQXMRSDOHW-UHFFFAOYSA-N
Synonym spermine, gerontine, musculamine, neuridine, spermin, 4,9-diaza-1,12-dodecanediamine, diaminopropyltetramethylenediamine, n,n'-bis 3-aminopropyl-1,4-butanediamine, spermine, puriss, 1,4-butanediamine, n,n'-bis 3-aminopropyl
PubChem CID 1103
ChEBI CHEBI:15746
IUPAC Name (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine
SMILES NCCCNCCCCNCCCN

Specifications

Color White
Quantity 1 g
Formula Weight 202.3
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Spermine
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.