Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermine Tetrahydrochloride, MP Biomedicals

Catalog No. MP210047410 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210047410 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210047410 Supplier MP Biomedicals, Inc Supplier No. 0210047410
Add to Cart
Add to Cart

  • Used to precipitate DNA from low salt aqueous buffers
  • Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors and red blood cells of chicken

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306-67-2
Molecular Formula C10H30Cl4N4
Molecular Weight (g/mol) 348.18
MDL Number MFCD00012914
InChI Key XLDKUDAXZWHPFH-UHFFFAOYSA-N
Synonym spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride
PubChem CID 9384
IUPAC Name tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride
SMILES [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN

Specifications

Melting Point 312°C to 314°C (literature)
Color White
Linear Formula C10H26N4 · 4HCl
Quantity 10 g
Solubility Information Soluble in water (100mg/mL - clear, colorless solution).
Formula Weight 348.2
Percent Purity ≥96%
Physical Form Powder
Chemical Name or Material Spermine Tetrahydrochloride
Recommended Storage -20°C, desiccate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.