CAS RN 306-67-2
Spermine tetrahydrochloride, 99%
CAS: 306-67-2 Molecular Formula: C10H30Cl4N4 Molecular Weight (g/mol): 348.18 MDL Number: MFCD00012914 InChI Key: XLDKUDAXZWHPFH-UHFFFAOYSA-N Synonym: spermine tetrahydrochloride,n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride,gerontine tetrahydrochloride,neuridine tetrahydrochloride,n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride,musculamine tetrahydrochloride,geontine tetrahydrochloride,spermine hcl,1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride,spermine, tetrahydrochloride PubChem CID: 9384 IUPAC Name: N,N'-bis(3-aminopropyl)butane-1,4-diamine;tetrahydrochloride SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN
Spermine Tetrahydrochloride 96-98% Crystalline MP Biomedicals
CAS: 306-67-2 Molecular Formula: C10H30Cl4N4 Molecular Weight (g/mol): 348.18 MDL Number: MFCD00012914 InChI Key: XLDKUDAXZWHPFH-UHFFFAOYSA-N Synonym: spermine tetrahydrochloride,n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride,gerontine tetrahydrochloride,neuridine tetrahydrochloride,n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride,musculamine tetrahydrochloride,geontine tetrahydrochloride,spermine hcl,1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride,spermine, tetrahydrochloride PubChem CID: 9384 IUPAC Name: tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN
Spermine Tetrahydrochloride, MP Biomedicals
CAS: 306-67-2 Molecular Formula: C10H30Cl4N4 Molecular Weight (g/mol): 348.18 MDL Number: MFCD00012914 InChI Key: XLDKUDAXZWHPFH-UHFFFAOYSA-N Synonym: spermine tetrahydrochloride,n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride,gerontine tetrahydrochloride,neuridine tetrahydrochloride,n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride,musculamine tetrahydrochloride,geontine tetrahydrochloride,spermine hcl,1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride,spermine, tetrahydrochloride PubChem CID: 9384 IUPAC Name: tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN
Spermine tetrahydrochloride, MP Biomedicals™
CAS: 306-67-2 Molecular Formula: C10H30Cl4N4 Molecular Weight (g/mol): 348.18 MDL Number: MFCD00012914 InChI Key: XLDKUDAXZWHPFH-UHFFFAOYSA-N Synonym: spermine tetrahydrochloride,n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride,gerontine tetrahydrochloride,neuridine tetrahydrochloride,n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride,musculamine tetrahydrochloride,geontine tetrahydrochloride,spermine hcl,1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride,spermine, tetrahydrochloride PubChem CID: 9384 IUPAC Name: tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN
N,N'-Bis(3-aminopropyl)-1,4-butanediamine Tetrahydrochloride 98.0+%, TCI America™
CAS: 306-67-2 Molecular Formula: C10H30Cl4N4 Molecular Weight (g/mol): 348.18 MDL Number: MFCD00012914 InChI Key: XLDKUDAXZWHPFH-UHFFFAOYSA-N Synonym: spermine tetrahydrochloride,n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride,gerontine tetrahydrochloride,neuridine tetrahydrochloride,n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride,musculamine tetrahydrochloride,geontine tetrahydrochloride,spermine hcl,1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride,spermine, tetrahydrochloride PubChem CID: 9384 IUPAC Name: tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN
R&D Systems™ Spermine tetrahydrochloride
CAS: 306-67-2 Molecular Formula: C10H30Cl4N4