Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spermine tetrahydrochloride, MP Biomedicals™

Catalog No. ICN10047401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10047401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10047401 Supplier MP Biomedicals, Inc Supplier No. 0210047401
Add to Cart
Add to Cart

Chemical Identifiers

CAS 306-67-2
Molecular Formula C10H30Cl4N4
Molecular Weight (g/mol) 348.18
MDL Number MFCD00012914
InChI Key XLDKUDAXZWHPFH-UHFFFAOYSA-N
Synonym spermine tetrahydrochloride, n,n'-bis 3-aminopropyl-1,4-butanediamine tetrahydrochloride, gerontine tetrahydrochloride, neuridine tetrahydrochloride, n1,n1'-butane-1,4-diyl bis propane-1,3-diamine tetrahydrochloride, musculamine tetrahydrochloride, geontine tetrahydrochloride, spermine hcl, 1,4-butanediamine, n,n'-bis 3-aminopropyl-, tetrahydrochloride, spermine, tetrahydrochloride
PubChem CID 9384
IUPAC Name tetrahydrogen (3-aminopropyl)({4-[(3-aminopropyl)amino]butyl})amine tetrachloride
SMILES [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCCNCCCCNCCCN

Specifications

Color White
Quantity 1 g
Formula Weight 348.19
Physical Form Solid
Chemical Name or Material Spermine tetrahydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.