Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEXQuE™ AOAC Internal Standard Mix, SPEX CertiPrep™
SDP

Catalog No. QUECRAOACIS Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QUECRAOACIS 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QUECRAOACIS Supplier SPEX CertiPrep Supplier No. QUECRAOACIS

missing translation for 'validMainframeMsg'

Sensible solutions for pesticide extraction

  • Solvent: Acetonitrile
  • Volume: 5mL

Specifications

Concentration 40 μg/mL
Technique Pesticide Extraction
Type AOAC Internal Standard Mix
Solvent or Matrix Acetonitrile
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.