Learn More
Description
Single Component Reference Standards
- Volume: 5mL
Specifications
Specifications
| Concentration | 10 μg/mL |
| For Use With (Application) | QuEChERS Analytical Techniques (GC, GC/MS, LC/MS) |
| Sorbent | Triphenylmethane |
| Type | QuEChERS standard- triphenylmethane |
| Volume (Metric) | 5 mL |
| Solvents | Acetonitrile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
