Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEX CertiPrep™ SPEXQuE™ QuEChERS Standards
SDP

Catalog No. QUECRPCB153 Shop All Spex CertiPrep Products
Change view
Click to view available options
Type:
QuEChERS standard- PCB 138
QuEChERS standard- PCB 153
QuEChERS standard- PCB 18
QuEChERS standard- PCB 52
QuEChERS standard- triphenylmethane
Sorbent:
PCB 138
PCB 153
PCB 18
PCB 52
Triphenylmethane
Concentration:
10 μg/mL
50 μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Sorbent Concentration
QUECRPCB153 QuEChERS standard- PCB 153 PCB 153 50 μg/mL
QUECRPCB138 QuEChERS standard- PCB 138 PCB 138 50 μg/mL
QUECRPCB18 QuEChERS standard- PCB 18 PCB 18 50 μg/mL
QUECRPCB52 QuEChERS standard- PCB 52 PCB 52 50 μg/mL
QUECR5190 QuEChERS standard- triphenylmethane Triphenylmethane 10 μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. QUECRPCB153 Supplier SPEX CertiPrep™ Supplier No. QUECRPCB153

SPEX CertiPrep provides a complete solution to all your QuEChERS needs with a comprehensive set of Internal Standards for the QuEChERS analytical techniques (GC, GC/MS, LC/MS)

Single Component Reference Standards

  • Volume: 5mL

Specifications

Concentration 50 μg/mL
For Use With (Application) QuEChERS Analytical Techniques (GC, GC/MS, LC/MS)
Sorbent PCB 153
Type QuEChERS standard- PCB 153
Volume (Metric) 5 mL
Solvents Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.