Learn More
Cole-Parmer™ Replacement Sample Extraction Tool for Microvial Grinding Set
Catalog No. 6753C
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
6753C 1 Ea.
1 options
Catalog No. 6753C Supplier Cole-Parmer™ Supplier No. 6753C
Add to Cart
Add to Cart
Sample Extraction Tool for removing ground samples from microvials which are used in the microvial set.
  • Insert the tool into an individual vial, then push it through to clear sample from the tube.
Specifications
Product Type Replacement Extraction Tool
For Use With (Equipment) 6757 Grinding Microvial Set (61043-06)
Packaging Quantity 1 Ea.