Learn More
SPEX™ SamplePrep Cryo-Adapter for 2210 Titer Plates
Catalog No. 2650
Change view
Click to view available options
For Use With (Equipment):
2210 Titer Plate
Catalog No. For Use With (Equipment)
2650 2210 Titer Plate
1 options
Catalog No. 2650 Supplier SPEX™ SamplePrep Supplier No. 2650
Add to Cart
Add to Cart
Extruded aluminum insert for 2210 96-well titer plate with round wells
  • Cryo-Blocks are chilled in the 2600 Cryo-Station and then used for high-throughput cryogenic grinding
  • Enables high-throughput cryogenic grinding in the Geno/Grinder™
  • Maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis
  • Preserves pesticides, RNA, and proteins for heat sensitive extraction methods
  • Increases yields by grinding samples at sub-zero temperatures
  • Used two at a time in the Standard Clamp Assembly to keep it in balance
Specifications
Product Type Cryo Adapter
For Use With (Application) Ideal for High Throughput Cryogenic Grinding
For Use With (Equipment) 2210 Titer Plate
Material Aluminum
WARNING: Reproductive Harm - www.P65Warnings.ca.gov