Learn More
Cole-Parmer™ Mid-Size Vial Adapter for Extractor 04576-38
Catalog No. 6884
Change view
Click to view available options
For Use With (Equipment):
6804 Large Extractor/Vial Opener (04576-38)
Catalog No. For Use With (Equipment)
68-84 6804 Large Extractor/Vial Opener (04576-38)
1 options
Catalog No. 68-84 Supplier Cole-Parmer™ Supplier No. 6884
Add to Cart
Add to Cart
Mid-Size Vial Adapter for use with SPEX SamplePrep™ Large-Capacity Freezer/Mill Cryogenic Grinder.
  • Converts Model 6804 for use with 6881, 6883 and 6885 grinding vial sets.
Specifications
Product Type Adapter
For Use With (Application) Converts 6804 for Use with 6881, 6883 and 6885 Grinding Vial Sets
For Use With (Equipment) 6804 Large Extractor/Vial Opener (04576-38)
WARNING: Cancer - www.P65Warnings.ca.gov