Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Grinding Vial Set, Large Vial, PC Cylinder, SS End Plugs and Impactor
GSA_VA

Catalog No. 6801
Change view
Click to view available options
Capacity (Metric):
1 to 50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
6801 1 to 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6801 Supplier Cole-Parmer™ Supplier No. 6801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Large Grinding Vial Set for use with SPEX SamplePrep™ Large-Capacity Freezer/Mill Cryogenic Grinder.

  • Makes one complete grinding vial plus three spare polycarbonate center cylinders.
  • Grinds one large sample at a time.
  • Sample capacity (grinding): 1 to 50 mL
  • Typical Sample Size: 25 g
  • Withstands liquid nitrogen temperatures.
  • Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples.
  • Transparent plastic center cylinder allows visual check of grinding process.
  • Stainless steel end plugs and impactor are autoclavable and reusable.
  • Polycarbonate center cylinders are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Compatible with 6804 Large Extractor/Vial Opener.
  • Components available separately.
  • For grinding samples such as plant and animal tissue, seeds, polymers, pharmaceuticals, textiles, hair, and bones.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Grinding Set
Capacity (English) 0.03 to 5.07 oz.
Capacity (Metric) 1 to 50 mL
For Use With (Application) Grinding Samples Such as Plant and Animal Tissue, Seeds, Polymers, Pharmaceuticals, Textiles, Hair and Bones
For Use With (Equipment) Large Extractor/Vial Opener 6804, Auto-Extractor 6814 of Spex SamplePrep Large Capacity Freezer/Mill
Includes 1 Large Stainless Steel Impactor, 2 Large Stainless Steel End Plugs, and 1 Pk. of Large Polycarbonate Center Cylinders
Material Stainless Steel
Material (Secondary) Polycarbonate
Sample Volume (English) 0.03 to 1.69 oz.
Sample Volume (Metric) 1 to 50 mL
Size Large
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.