Learn More
Cole-Parmer™ Replacement Cylinder for Large Grinding Vial Sets
GSA_VA
Catalog No. 6802
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
68-02 1 Ea.
1 options
Catalog No. 68-02 Supplier Cole-Parmer™ Supplier No. 6802
Add to Cart
Add to Cart
Alternative stainless steel center cylinder for 6801 Large Grinding Vial Set.
  • Replaces polycarbonate center cylinders when sample contact with plastic is not acceptable.
  • Made of nonmagnetic 440C stainless steel.
  • Withstands liquid nitrogen temperatures.
  • Autoclavable and reusable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Center Cylinder
Autoclavable Autoclavable
Disposable Reusable
For Use With (Application) Replaces Polycarbonate Cylinder When Sample Contact with Plastic Cannot be Used
For Use With (Equipment) 6801 Large Grinding Vial Set (61043-10)
Material Stainless Steel (440 C)
Packaging Quantity 1 Ea.