Learn More
Cole-Parmer™ Replacement End Plugs for Mid-Size Grinding Vial Set, Stainless Steel
Catalog No. 6881E
Change view
Click to view available options
For Use With (Equipment):
6881 Mid Size Grinding Vial Set (61043-08)
Catalog No. For Use With (Equipment)
68-81E 6881 Mid Size Grinding Vial Set (61043-08)
1 options
Catalog No. 68-81E Supplier Cole-Parmer™ Supplier No. 6881E
Add to Cart
Add to Cart
Replacement mid-size stainless-steel end plugs for 6881 Mid-Size Grinding Vial Set
  • Made of 440C stainless steel.
  • Withstands liquid nitrogen temperatures.
  • Autoclavable and reusable.
Specifications
Product Type End Plug
Disposable Reusable
For Use With (Equipment) 6881 Mid Size Grinding Vial Set (61043-08)
Material Stainless Steel (440 C)
Packaging Quantity 2 Pk.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov