Learn More
Cole-Parmer™ Grinding Ball Dispenser for 96-Well Microplates
Catalog No. 2100
Change view
Click to view available options
For Use With (Equipment):
Grinding Ball and 96-Well Titer Plates of Geno/Grinder
Catalog No. For Use With (Equipment)
2100 Grinding Ball and 96-Well Titer Plates of Geno/Grinder
1 options
Catalog No. 2100 Supplier Cole-Parmer™ Supplier No. 2100
Add to Cart
Add to Cart
Grinding Ball Dispenser for use with Geno/Grinder™.
  • Loads each well of a standard 96-well titer plate with one grinding ball in less than a second.
Specifications
Product Type Grinding Ball Dispenser
For Use With (Equipment) Grinding Ball and 96-Well Titer Plates of Geno/Grinder
Holds 96-well Microplates
Material Stainless Steel (440 C)
Packaging Quantity 1 Ea.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov