Learn More
New Pig Sphergo Swivel™ Disposable Cleanroom Cleaning Pad
SDP
Catalog No. 17987827 Shop All New Pig Corporation Products
Catalog No. 17-987-827 Supplier New Pig Supplier No. CLN381
Add to Cart
Add to Cart
Mop your cleanroom with these disposable cleaning pads that last over a larger surface area
  • Single-use design for contamination control: These pads are disposable, ensuring that contaminants are not carried from one area of your cleanroom to another.
  • Superior absorbency and fluid release: The microfiber material absorbs up to 9 times its weight in liquid, while also releasing it consistently over a large surface area for efficient cleaning.
  • Low-lint and high-performance: Bonded edges and a specialized design reduce linting, while an embossed pattern minimizes friction for easier, more effective cleaning.
  • Ideal for critical environments: Compatible with common disinfectants and formulated for cleanroom use, these pads meet strict standards such as USP 797 compliance.
  • Ergonomically designed for reduced strain: When used with the Sphergo Swivel™ mop system, these pads allow for smooth, effortless S motion cleaning, reducing worker fatigue.
  • Eco-friendly materials: Made with 50% post-consumer recycled content, these pads offer a sustainable option for your cleanroom cleaning needs.