Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Spherical Pinch Clamp
GREENER_CHOICESDP

Catalog No. 31501931 Shop All Kemtech Products
Change view
Click to view available options
Size:
12/5, #5
18/8, #9
28/15, #15
35/20, #25
50/30, #30
65/40, #40
75/50, #50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
31-501-931 18/8, #9
31-501-930 12/5, #5
31-501-932 28/15, #15
31-501-933 35/20, #25
31-501-934 50/30, #30
31-501-935 65/40, #40
31-501-936 75/50, #50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 31-501-931 Supplier Synthware™ Supplier No. CC0018
Add to Cart
Add to Cart

Spherical Pinch Clamp features 304 Stainless Steel construction with a screw type lock nut.

  • With a screw type locking nut.

Specifications

Type Pinch Clamp
Material Stainless Steel
Includes Screw Down Locking Nut
For Use With (Application) Organic Synthesis
Size 18/8, #9
Color Silver
Jaw Type Spring Loaded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.