Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Sphingomyelin Assay Kit, Fluorometric, Quantitative, 100 Tests
Description
- Detection method: Fluorescent
- Sensitivity: 1 μM
- Sample types: Cell culture extracts, Whole blood
- Results type: Quantitative
- Application: Measure sphingomyelin(43:7) in cell/tissue extracts and biofluids
- Instrumentation: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Sphingomyelin |
| Substrate Properties | Lipid-based Substrate |
| Detection Method | Fluorescence |
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 100 Tests |
| Sample Type | Cell culture extracts, Whole blood |
| Quantity | 100 Tests |
| For Use With (Application) | Sphingomyelin assay |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?