Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sphingosine-1-phosphate
GSA_VAGREENER_CHOICESDP

Catalog No. 13701
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-701 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-701 Supplier Tocris Bioscience™ Supplier No. 1370/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Endogenous agonist at S1P1-5

Endogenous second messenger involved in the control of cell proliferation and motility, and Ca2+ mobilization. Acts as an agonist at sphingosine-1-phosphate receptors (S1P1-5) and as an activator of GPR3, GPR6 and GPR12. Effectors regulated include p38 MAP kinase, PLC, adenylyl cyclase, myosin light chain phosphatase and focal adhesion kinase.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 26993-30-6
Molecular Formula C18H38NO5P

Specifications

Quantity 1 mg
Formula Weight 379.48
Purity >98%
Molecular Weight (g/mol) 379.478
Chemical Name or Material Sphingosine-1-phosphate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.