Learn More
Echelon Biosciences Research Labs D-Erythro-Sphingosine 25 mg
SDP
Supplier:  Echelon Biosciences Research Labs S100025MG
Encompass
Sphingosine is formed through ceramidase catalyzed hydrolysis of ceramide. It is a potent inhibitor of protein kinase C (IC50 = 1-3 M). Sphingosine is phosphorylated by sphingosine kinase to the bioactive lipid sphingosine 1-phosphate This item is non returnable
Catalog No. 50-720-763
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.