Learn More
Echelon Biosciences Research Labs SPHINGOSINE-BIOTIN 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs S100B50UG
Encompass
Echelon offers a unique selection of sphingosine analogs labeled at the terminus of the alkyl chain with biotin or fluorescent groups. All compounds are synthesized from chiral precursors and are available as lyophilized powders. Echelons alkyl-labeled sphingosines are synthesized with the natural erythro stereochemistry and maintain similar properties as sphingosine. For example, labeled sphingosines are phosphorylated by sphingosine kinases (1 & 2) and can be used in high throughput screening applications. In addition, they induce a Nieman Pick type C1 disease cellular phenoype in RAW macrophages in the same manner as exogenously added sphingosine. This item is non returnable
Catalog No. 50-720-764
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.