Learn More
Enzo Life Sciences Sphingosine, D-erythro (25mg). CAS: 123-78-4
SDP
Supplier:  Enzo Life Sciences BMLEI1550025
Encompass_Preferred
Common component of cell membranes, often as phosphatidyl conjugates. Is a potent (IC50=1-3 µM) and selective inhibitor of protein kinase C. Inhibition is competitive with diacylglycerol, phorbol dibutyrate and Ca2+ and it also inhibits PKC activation by other lipids. It is active in intact cells and has been identified as an endogenous constituent in HL-60 cells, neutrophils, rat liver, and brain and mouse tissues. Other activities include inhibition of phosphatidate phosphohydrolase, Na+,K+-ATPase, CTP:phosphocholine cytidylyltransferase, calmodulin-dependent enzymes, binding of factor VII to tissue factor, binding of thyrotropin releasing hormone to its receptor and activation of EGF receptor kinase, phospholipase D and casein kinase II. Alternative name: D(+)-erythro-1,3-Dihydroxy-2-amino-4- trans-octadecene. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol (25mg/ml warm) or DMSO (25mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-0577
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.